Gene Transcripts

Gene Symbol
NOC4L
Entrez Gene ID
79050
Gene Name
nucleolar complex associated 4 homolog
Gene Aliases
NET49, NOC4, UTP19
Chromosome Location
Chr. 12: 132144421 - 132152473 on Build GRCh38
UniGene ID
Hs.558536
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024078.2 NP_076983.1 10 1006
XM_017019960.1 XP_016875449.1 9 960
GenBank mRNA BC001191.1 - 10 985
BC007893.2 - 10 878
BC071958.1 - 10 949 6597539

View Full Product Details