Gene Transcripts
- Gene Symbol
- PHF23
- Entrez Gene ID
- 79142
- Gene Name
- PHD finger protein 23
- Gene Aliases
- hJUNE-1b
- Chromosome Location
- Chr. 17: 7235028 - 7239506 on Build GRCh38
- UniGene ID
- Hs.644724
- Species
- Homo sapiens
- Gene Symbol
- NUP98
- Entrez Gene ID
- 4928
- Gene Name
- nucleoporin 98
- Gene Aliases
- ADIR2, NUP196, NUP96
- Chromosome Location
- Chr. 11: 3675010 - 3797792 on Build GRCh38
- UniGene ID
- Hs.524750
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
KU847959.1 |
- |
|
1648 |
|
View Full Product Details