Gene Transcripts

Gene Symbol
C10orf76
Entrez Gene ID
79591
Gene Name
chromosome 10 open reading frame 76
Gene Aliases
Chromosome Location
Chr. 10: 101845599 - 102056237 on Build GRCh38
UniGene ID
Hs.16004
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024541.2 NP_078817.2 21 1624
XM_011540151.2 XP_011538453.1 21 1669
XM_011540152.2 XP_011538454.1 20 1556
XM_011540153.2 XP_011538455.1 20 1520
XM_011540154.2 XP_011538456.1 19 1454
XM_011540156.2 XP_011538458.1 14 1588
XM_011540157.2 XP_011538459.1 20 1573
XM_017016635.1 XP_016872124.1 21 1622
XM_017016636.1 XP_016872125.1 21 1622
GenBank mRNA AK023176.1 - 4 253
BC110511.1 - 4 253

View Full Product Details