Gene Transcripts

Gene Symbol
HECTD3
Entrez Gene ID
79654
Gene Name
HECT domain E3 ubiquitin protein ligase 3
Gene Aliases
Chromosome Location
Chr. 1: 45002548 - 45011355 on Build GRCh38
UniGene ID
Hs.525084
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024602.5 NP_078878.3 16 2118
XM_011542140.2 XP_011540442.1 14 1515
GenBank mRNA AK056545.1 - 14 1469
AK056873.1 - 8 1012
AK091583.1 - 1 867
AK096462.1 - 16 1997
AK301124.1 - 15 1706
BC019105.2 - 1 77
BC126352.1 - 13 1395 8992068
BC130470.1 - 13 1395 40146501

View Full Product Details