Gene Transcripts

Gene Symbol
VEPH1
Entrez Gene ID
79674
Gene Name
ventricular zone expressed PH domain containing 1
Gene Aliases
MELT, VEPH
Chromosome Location
Chr. 3: 157259742 - 157503626 on Build GRCh38
UniGene ID
Hs.658046
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167911.1 NP_001161383.1 6 975
NM_001167912.1 NP_001161384.1 6 975
NM_024621.2 NP_078897.2 6 1166
XM_011513134.2 XP_011511436.1 5 909
XM_011513135.1 XP_011511437.1 6 805
GenBank mRNA AB051479.1 - 6 975
AK027625.1 - 6 1064
AL713656.1 - 5 763
BC101660.1 - 5 802 8069166
BC111017.1 - 6 967 5174889
BC113555.1 - 5 802 8322607

View Full Product Details