Gene Transcripts
- Gene Symbol
- CENPU
- Entrez Gene ID
- 79682
- Gene Name
- centromere protein U
- Gene Aliases
- CENP50, CENPU50, KLIP1, MLF1IP, PBIP1
- Chromosome Location
- Chr. 4: 184694065 - 184734132 on Build GRCh38
- UniGene ID
- Hs.575032
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024629.3 |
NP_078905.2
|
12, 13 |
1205 |
|
NR_104593.1 |
|
11, 12 |
1143 |
|
XM_005263218.4 |
XP_005263275.2
|
12, 13 |
1240 |
|
GenBank mRNA |
AF469667.1 |
- |
12, 13 |
1208 |
|
AF516710.1 |
- |
13, 14 |
1622 |
|
AK027121.1 |
- |
10, 11 |
1015 |
|
AK292297.1 |
- |
12, 13 |
1151 |
|
BC031520.1 |
- |
7, 8 |
553 |
|
BC067802.1 |
- |
11, 12 |
1083 |
|
BC107744.1 |
- |
7, 8 |
650 |
|
BC131556.1 |
- |
12, 13 |
1205 |
40108113 |
BC141854.1 |
- |
11, 12 |
1141 |
8860370 |
BG031878.1 |
- |
1, 2 |
47 |
|
CR457376.1 |
- |
5, 6 |
409 |
|
View Full Product Details