Gene Transcripts

Gene Symbol
ZDHHC11B
Entrez Gene ID
653082
Gene Name
zinc finger DHHC-type containing 11B
Gene Aliases
DHHC-11B
Chromosome Location
Chr. 5: 710355 - 784749 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017010110.1 XP_016865599.1 5 582
XM_017010111.1 XP_016865600.1 4 409
XM_017010112.1 XP_016865601.1 4 986
XM_017010113.1 XP_016865602.1 5 1108
XM_017010114.1 XP_016865603.1 5 3869
XM_017010115.1 XP_016865604.1 4 934
XM_017010116.1 XP_016865605.1 3 1334
XM_017010117.1 XP_016865606.1 3 1334
XM_017010118.1 XP_016865607.1 3 1334
XM_017010119.1 XP_016865608.1 3 1334
XM_017010120.1 XP_016865609.1 3 1334
XM_017010121.1 XP_016865610.1 3 1334
XM_017010122.1 XP_016865611.1 3 1334
XM_017010123.1 XP_016865612.1 3 1334
XM_017010124.1 XP_016865613.1 5 582

Gene Symbol
ZDHHC11
Entrez Gene ID
79844
Gene Name
zinc finger DHHC-type containing 11
Gene Aliases
ZNF399
Chromosome Location
Chr. 5: 795605 - 863634 on Build GRCh38
UniGene ID
Hs.654874
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024786.2 NP_079062.1 1 482
XM_017009869.1 XP_016865358.1 2 198
XM_017009870.1 XP_016865359.1 4 2009
XM_017009871.1 XP_016865360.1 4 1958
XM_017009872.1 XP_016865361.1 2 198
GenBank mRNA AK023215.1 - 1 482
BC032000.1 - 1 101 4750082

View Full Product Details