Gene Transcripts
- Gene Symbol
- TTC26
- Entrez Gene ID
- 79989
- Gene Name
- tetratricopeptide repeat domain 26
- Gene Aliases
- DYF13, IFT56, dyf-13
- Chromosome Location
- Chr. 7: 139133744 - 139191986 on Build GRCh38
- UniGene ID
- Hs.133020
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK022633.1 |
- |
16 |
1490 |
|
View Full Product Details