Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007038.4 | NP_008969.2 | 8 | 3125 | |
XM_011529448.2 | XP_011527750.1 | 7 | 2957 | ||
GenBank mRNA | AF141293.1 | - | 6 | 1160 | |
AF142099.1 | - | 8 | 2518 | ||
BC093775.1 | - | 8 | 2480 | 7939620 | |
BC093777.1 | - | 8 | 2480 | 7939622 |