Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286496.1 | NP_001273425.1 | 4 | 830 | |
NM_001286497.1 | NP_001273426.1 | 4 | 806 | ||
NM_001286499.1 | NP_001273428.1 | 4 | 830 | ||
NM_025049.3 | NP_079325.2 | 4 | 806 | ||
XM_011522083.2 | XP_011520385.1 | 4 | 922 | ||
XM_011522084.2 | XP_011520386.1 | 4 | 754 | ||
GenBank mRNA | AB185926.1 | - | 4 | 806 | |
AB185927.1 | - | 4 | 806 | ||
AK026345.1 | - | 3 | 849 | ||
AY498716.1 | - | 3 | 730 | ||
BC137503.1 | - | 4 | 744 | 9021503 | |
BC137504.1 | - | 4 | 744 | 9021504 | |
DQ437529.1 | - | 3 | 711 | ||
EU084033.1 | - | 4 | 801 |