Gene Transcripts

Gene Symbol
WDR26
Entrez Gene ID
80232
Gene Name
WD repeat domain 26
Gene Aliases
CDW2, GID7, MIP2
Chromosome Location
Chr. 1: 224385143 - 224434299 on Build GRCh38
UniGene ID
Hs.497873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001115113.2 NP_001108585.2 9, 10 1568
NM_025160.6 NP_079436.4 9, 10 1616
XM_005273273.4 XP_005273330.1 9, 10 1517
XM_006711817.3 XP_006711880.1 9, 10 1514
XM_011544281.2 XP_011542583.1 9, 10 1357
XM_011544282.2 XP_011542584.1 6, 7 679
XM_017002396.1 XP_016857885.1 9, 10 1205
GenBank mRNA AB209887.1 - 9, 10 1616
AK024669.1 - 5, 6 630
AK225061.1 - 5, 6 630
AK299230.1 - 6, 7 801
AY221751.1 - 9, 10 1406
AY304473.1 - 9, 10 1173
BC031471.2 - 4, 5 559
BC034498.1 - 11, 12 1608
BC052301.1 - 8, 9 1012 6420398
BC063817.1 - 5, 6 678
EF011612.1 - 8, 9 1011

View Full Product Details