Gene Transcripts

Gene Symbol
WDCP
Entrez Gene ID
80304
Gene Name
WD repeat and coiled coil containing
Gene Aliases
C2orf44, PP384
Chromosome Location
Chr. 2: 24029336 - 24047426 on Build GRCh38
UniGene ID
Hs.24624
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142319.1 NP_001135791.1 2 1118
NM_025203.2 NP_079479.1 2 1118
XM_017005029.1 XP_016860518.1 2 1113
GenBank mRNA AK025598.1 - 2 1045
BC035698.2 - 2 1053 5547301

View Full Product Details