Gene Transcripts

Gene Symbol
KHDC1
Entrez Gene ID
80759
Gene Name
KH homology domain containing 1
Gene Aliases
C6orf147, C6orf148, Em:AC019205.8, NDG1, bA257K9.4
Chromosome Location
Chr. 6: 73241314 - 73310365 on Build GRCh38
UniGene ID
Hs.433062
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001251874.1 NP_001238803.1 5 1185
NM_030568.4 NP_085045.3 4 901
GenBank mRNA BC022080.2 - 4 871
BM725230.1 - 3 444

View Full Product Details