Gene Transcripts

Gene Symbol
PRRT1
Entrez Gene ID
80863
Gene Name
proline rich transmembrane protein 1
Gene Aliases
C6orf31, DSPD1, IFITMD7, NG5
Chromosome Location
Chr. 6: 32148363 - 32151943 on Build GRCh38
UniGene ID
Hs.549204
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030651.3 NP_085154.3 2 259
GenBank mRNA AK123424.1 - 1 1141
BC063046.1 - 2 207 5197474

View Full Product Details