Gene Transcripts

Gene Symbol
OR8J3
Entrez Gene ID
81168
Gene Name
olfactory receptor family 8 subfamily J member 3
Gene Aliases
OR11-173
Chromosome Location
Chr. 11: 56136771 - 56137718 on Build GRCh38
UniGene ID
Hs.554523
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004064.1 NP_001004064.1 1 615

View Full Product Details