Gene Transcripts

Gene Symbol
PTDSS2
Entrez Gene ID
81490
Gene Name
phosphatidylserine synthase 2
Gene Aliases
PSS2
Chromosome Location
Chr. 11: 448264 - 491399 on Build GRCh38
UniGene ID
Hs.731700
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030783.1 NP_110410.1 8 1020
XM_005253153.1 XP_005253210.1 8 1001
XM_011520391.2 XP_011518693.1 8 1638
XM_011520392.2 XP_011518694.1 9 1017
XM_011520393.2 XP_011518695.1 7 735
XM_017018368.1 XP_016873857.1 10 3412
XM_017018369.1 XP_016873858.1 9 1142
XM_017018370.1 XP_016873859.1 7 817
XM_017018371.1 XP_016873860.1 8 931
GenBank mRNA AL834357.1 - 8 934
BC001210.1 - 8 1020 3503123

View Full Product Details