Gene Transcripts

Gene Symbol
C1orf21
Entrez Gene ID
81563
Gene Name
chromosome 1 open reading frame 21
Gene Aliases
PIG13
Chromosome Location
Chr. 1: 184387016 - 184629021 on Build GRCh38
UniGene ID
Hs.497159
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030806.3 NP_110433.1 2, 3 578
GenBank mRNA AF312864.1 - 2, 3 500
AK056487.1 - 2, 3 314
AK312062.1 - 1, 2 215
AY258286.1 - 2, 3 491
BC028567.2 - 2, 3 536 3635521
BG823539.1 - 2, 3 568

View Full Product Details