Gene Transcripts

Gene Symbol
ANKRD13C
Entrez Gene ID
81573
Gene Name
ankyrin repeat domain 13C
Gene Aliases
dJ677H15.3
Chromosome Location
Chr. 1: 70259002 - 70354977 on Build GRCh38
UniGene ID
Hs.744989
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030816.4 NP_110443.3 1, 2 748
XM_005271234.3 XP_005271291.1 1, 2 991
XM_005271235.3 XP_005271292.1 1, 2 738
XM_006710929.3 XP_006710992.1 1, 2 761
XM_017002414.1 XP_016857903.1 1, 2 99
XM_017002415.1 XP_016857904.1 1, 2 99
GenBank mRNA AL136717.1 - 1, 2 749
BC028840.1 - 1, 2 737 4558748

View Full Product Details