Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001202543.1 | NP_001189472.1 | 2 | 260 | |
NM_001202544.2 | NP_001189473.1 | 2 | 260 | ||
NM_001202545.2 | NP_001189474.1 | 2 | 260 | ||
NM_001913.4 | NP_001904.2 | 2 | 260 | ||
NM_181500.3 | NP_852477.1 | 2 | 260 | ||
NM_181552.3 | NP_853530.2 | 2 | 138 | ||
XM_005250150.2 | XP_005250207.1 | 2 | 659 | ||
XM_005250151.2 | XP_005250208.1 | 2 | 662 | ||
XM_005250154.3 | XP_005250211.1 | 2 | 516 | ||
XM_006715854.2 | XP_006715917.1 | 2 | 659 | ||
XM_006715855.1 | XP_006715918.1 | 2 | 516 | ||
XM_011515823.2 | XP_011514125.1 | 2 | 657 | ||
XM_011515825.1 | XP_011514127.1 | 2 | 225 | ||
XM_017011759.1 | XP_016867248.1 | 2 | 225 | ||
XM_017011760.1 | XP_016867249.1 | 2 | 225 | ||
GenBank mRNA | AK122726.1 | - | 2 | 143 | |
AK125097.1 | - | 2 | 155 | ||
AK222832.1 | - | 2 | 156 | ||
AK297548.1 | - | 2 | 146 | ||
BC025422.2 | - | 2 | 141 | ||
BC066592.1 | - | 2 | 153 | 5740343 | |
DC410774.1 | - | 2 | 260 | ||
L12579.1 | - | 2 | 152 | ||
M74099.1 | - | 2 | 143 |