Gene Transcripts

Gene Symbol
URM1
Entrez Gene ID
81605
Gene Name
ubiquitin related modifier 1
Gene Aliases
C9orf74
Chromosome Location
Chr. 9: 128371319 - 128392016 on Build GRCh38
UniGene ID
Hs.495229
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135947.2 NP_001129419.1 2 138
NM_001265582.1 NP_001252511.1 2 138
NM_030914.3 NP_112176.1 2 138
NR_049743.1 2 179
GenBank mRNA AK001880.1 - 2 87
AK097029.1 - 2 84
AK300877.1 - 2 85
BC003581.1 - 2 96 3546655
BI823604.1 - 2 138
D83200.1 - 1 44
DA691216.1 - 2 144

View Full Product Details