Gene Transcripts

Gene Symbol
C6orf62
Entrez Gene ID
81688
Gene Name
chromosome 6 open reading frame 62
Gene Aliases
Nbla00237, XTP12, dJ30M3.2
Chromosome Location
Chr. 6: 24704758 - 24720836 on Build GRCh38
UniGene ID
Hs.744857
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030939.4 NP_112201.1 2 795
XM_005249433.3 XP_005249490.1 2 1202
XM_011514928.2 XP_011513230.1 2 1136
GenBank mRNA AB075502.1 - 2 782
AK022681.1 - 2 679
AK301718.1 - 2 334
AL136632.2 - 2 806
AY598792.1 - 2 288
BC040260.1 - 2 307
BC047866.1 - 2 486 6499979
CR457292.1 - 2 288
CR533486.1 - 2 288

View Full Product Details