Gene Transcripts

Gene Symbol
KRTAP1-3
Entrez Gene ID
81850
Gene Name
keratin associated protein 1-3
Gene Aliases
KAP1.2, KAP1.3, KAP1.6, KAP1.8A, KAP1.8B, KAP1.9, KRTAP1.3
Chromosome Location
Chr. 17: 41033884 - 41034855 on Build GRCh38
UniGene ID
Hs.534495
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030966.1 NP_112228.1 1 945
GenBank mRNA AJ406927.1 - 1 945
BC069161.1 - 1 944 7216931

View Full Product Details