Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031430.2 | NP_113618.2 | 5 | 1061 | |
XM_005256811.1 | XP_005256868.1 | 5 | 1040 | ||
GenBank mRNA | AF370391.1 | - | 5 | 998 | |
AJ278711.1 | - | 5 | 1061 | ||
AJ404317.1 | - | 5 | 1031 | ||
AK055755.1 | - | 5 | 1040 | ||
AK314767.1 | - | 5 | 1040 | ||
BC004961.1 | - | 5 | 610 | ||
BC031621.1 | - | 5 | 855 | 5174974 |