Gene Transcripts

Gene Symbol
SLC35G5
Entrez Gene ID
83650
Gene Name
solute carrier family 35 member G5
Gene Aliases
AMAC, AMAC1L2
Chromosome Location
Chr. 8: 11330986 - 11332186 on Build GRCh38
UniGene ID
Hs.458397
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054028.1 NP_473369.1 1 531
GenBank mRNA AJ291677.1 - 1 531
BC131696.1 - 1 498 40120574

View Full Product Details