Gene Transcripts

Gene Symbol
TEKT1
Entrez Gene ID
83659
Gene Name
tektin 1
Gene Aliases
Chromosome Location
Chr. 17: 6799981 - 6831741 on Build GRCh38
UniGene ID
Hs.462108
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053285.1 NP_444515.1 3 381
XM_011524027.2 XP_011522329.1 3 377
GenBank mRNA AF357879.1 - 3 330
AK292818.1 - 3 346
BC014599.2 - 3 381 4824860

View Full Product Details