Gene Transcripts

Gene Symbol
TBC1D10A
Entrez Gene ID
83874
Gene Name
TBC1 domain family member 10A
Gene Aliases
EPI64, TBC1D10, dJ130H16.1, dJ130H16.2
Chromosome Location
Chr. 22: 30291990 - 30326966 on Build GRCh38
UniGene ID
Hs.655273
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204240.1 NP_001191169.1 2 381
NM_031937.2 NP_114143.1 2 360
GenBank mRNA AB449894.1 - 2 275
AF331038.1 - 2 299
AK074656.1 - 2 326
AK127939.1 - 2 316
AK309455.1 - 2 126
BC022201.1 - 2 309
BC136815.1 - 2 316 9020806
BC142940.1 - 2 332 8860306
BC150187.1 - 2 280 40148357
BC150214.1 - 2 285 40149007
BC171775.1 - 2 316 9054429
CT841514.1 - 2 281
DA944522.1 - 2 363

View Full Product Details