Gene Transcripts
- Gene Symbol
- MND1
- Entrez Gene ID
- 84057
- Gene Name
- meiotic nuclear divisions 1
- Gene Aliases
- GAJ
- Chromosome Location
- Chr. 4: 153344649 - 153415097 on Build GRCh38
- UniGene ID
- Hs.294088
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001253861.1 |
NP_001240790.1
|
6, 7 |
483 |
|
NM_032117.3 |
NP_115493.1
|
7, 8 |
598 |
|
NR_045605.1 |
|
9, 10 |
759 |
|
XM_017008688.1 |
XP_016864177.1
|
7, 8 |
955 |
|
GenBank mRNA |
AK312172.1 |
- |
7, 8 |
557 |
|
AY028916.1 |
- |
7, 8 |
537 |
|
BC032142.2 |
- |
7, 8 |
552 |
5095005 |
BG613292.1 |
- |
6, 7 |
445 |
|
BG758047.1 |
- |
9, 10 |
728 |
|
BU533354.1 |
- |
4, 5 |
254 |
|
View Full Product Details