Gene Transcripts

Gene Symbol
KIRREL2
Entrez Gene ID
84063
Gene Name
kin of IRRE like 2 (Drosophila)
Gene Aliases
FILTRIN, NEPH3, NLG1
Chromosome Location
Chr. 19: 35851635 - 35867146 on Build GRCh38
UniGene ID
Hs.145729
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032123.6 NP_115499.5 16 1930
NM_199179.3 NP_954648.3 15 1780
NM_199180.3 NP_954649.3 15 2341
XM_011527362.1 XP_011525664.1 16 2401
XM_011527363.1 XP_011525665.1 16 2382
XM_011527365.1 XP_011525667.1 15 2251
XM_011527366.1 XP_011525668.1 14 2512
XM_017027348.1 XP_016882837.1 14 2803
GenBank mRNA AB593116.1 - 15 2305
AL136654.1 - 15 2325
AY305301.1 - 16 1874
AY305302.1 - 15 1724
AY358742.1 - 15 2327
BC007312.1 - 6 874
BC064925.1 - 18 2264 6141879

View Full Product Details