Gene Transcripts

Gene Symbol
ENKD1
Entrez Gene ID
84080
Gene Name
enkurin domain containing 1
Gene Aliases
C16orf48, DAKV6410
Chromosome Location
Chr. 16: 67662947 - 67666725 on Build GRCh38
UniGene ID
Hs.729159
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032140.1 NP_115516.1 3 611
GenBank mRNA AL136786.1 - 3 611
BC008284.1 - 3 578 3638194

View Full Product Details