Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168378.1 | NP_001161850.1 | 3 | 643 | |
NM_001168379.1 | NP_001161851.1 | 3 | 470 | ||
NM_032153.5 | NP_115529.2 | 3 | 769 | ||
XM_017007314.1 | XP_016862803.1 | 3 | 3252 | ||
GenBank mRNA | AF332509.1 | - | 1 | 1162 | |
AK056262.1 | - | 3 | 580 | ||
AK056394.1 | - | 1 | 954 | ||
AK094286.1 | - | 1 | 266 | ||
AK294831.1 | - | 3 | 643 | ||
BC029507.1 | - | 3 | 548 | ||
BC126267.1 | - | 3 | 405 | 8991983 | |
BC136339.1 | - | 3 | 405 | 9020326 | |
DA066220.1 | - | 3 | 498 |