Gene Transcripts

Gene Symbol
SARNP
Entrez Gene ID
84324
Gene Name
SAP domain containing ribonucleoprotein
Gene Aliases
CIP29, HCC1, HSPC316, THO1
Chromosome Location
Chr. 12: 55752463 - 55817756 on Build GRCh38
UniGene ID
Hs.505676
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033082.3 NP_149073.1 2 148
NR_026722.1 2 148
NR_026723.1 2 148
GenBank mRNA AF161434.1 - 2 115
AF486281.1 - 2 93
AK222719.1 - 2 126
AK290508.1 - 2 116
BC007099.1 - 2 114 4273903
BC093051.1 - 2 114 30382893
BI520421.1 - 2 148

View Full Product Details