Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195483.1 | NP_001182412.1 | 9 | 1569 | |
NM_024628.5 | NP_078904.3 | 10 | 1630 | ||
GenBank mRNA | AF345197.1 | - | 9 | 1519 | |
AF390442.1 | - | 9 | 1570 | ||
AK026841.1 | - | 6 | 966 | ||
AK123001.1 | - | 10 | 1584 | ||
AK308484.1 | - | 5 | 887 | ||
BC020506.1 | - | 2 | 532 | ||
BC063528.1 | - | 5 | 853 | ||
BC126158.1 | - | 9 | 1540 | 8991874 | |
BC126160.1 | - | 9 | 1540 | 8991876 | |
BX538023.1 | - | 4 | 953 | ||
CR457363.1 | - | 1 | 301 |