Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001195483.1 | NP_001182412.1 | 9 | 1727 | |
NM_024628.5 | NP_078904.3 | 10 | 1788 | ||
GenBank mRNA | AF345197.1 | - | 9 | 1677 | |
AF390442.1 | - | 9 | 1728 | ||
AK026841.1 | - | 6 | 1124 | ||
AK123001.1 | - | 10 | 1742 | ||
AK308484.1 | - | 5 | 1045 | ||
BC020506.1 | - | 2 | 690 | ||
BC063528.1 | - | 5 | 1011 | ||
BC126158.1 | - | 9 | 1698 | 8991874 | |
BC126160.1 | - | 9 | 1698 | 8991876 | |
BX538023.1 | - | 4 | 1111 | ||
CR457363.1 | - | 1 | 459 |