Gene Transcripts

Gene Symbol
KRTAP4-4
Entrez Gene ID
84616
Gene Name
keratin associated protein 4-4
Gene Aliases
KAP4.13, KAP4.4, KRTAP4-13, KRTAP4.13, KRTAP4.4
Chromosome Location
Chr. 17: 41159653 - 41160731 on Build GRCh38
UniGene ID
Hs.307022
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032524.1 NP_115913.1 1 922
GenBank mRNA AJ296168.1 - 2 745
AJ406936.1 - 1 922

View Full Product Details