Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289158.1 | NP_001276087.1 | 5 | 505 | |
NM_032571.4 | NP_115960.2 | 6 | 661 | ||
XM_011528374.2 | XP_011526676.1 | 5 | 552 | ||
GenBank mRNA | AF239764.1 | - | 6 | 598 | |
AK290909.1 | - | 6 | 585 | ||
AL832663.1 | - | 6 | 662 | ||
AY358817.1 | - | 5 | 402 | ||
BC121130.2 | - | 6 | 583 | ||
BC121131.2 | - | 6 | 583 | ||
BC128053.1 | - | 6 | 583 | ||
BC148282.2 | - | 6 | 583 |