Gene Transcripts
- Gene Symbol
- TXLNGY
- Entrez Gene ID
- 246126
- Gene Name
- taxilin gamma pseudogene, Y-linked
- Gene Aliases
- CYorf15A, CYorf15B, TXLNG2P
- Chromosome Location
- Chr. Y: 19567358 - 19607170 on Build GRCh38
- UniGene ID
- Hs.522863
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045128.1 |
|
10 |
1545 |
|
GenBank mRNA |
AF332225.1 |
- |
4 |
821 |
|
AL832583.2 |
- |
5 |
5091 |
|
View Full Product Details