Gene Transcripts

Gene Symbol
KDM2B
Entrez Gene ID
84678
Gene Name
lysine demethylase 2B
Gene Aliases
CXXC2, FBXL10, Fbl10, JHDM1B, PCCX2
Chromosome Location
Chr. 12: 121402289 - 121582268 on Build GRCh38
UniGene ID
Hs.524800
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005366.1 NP_001005366.1 22 4041
NM_032590.4 NP_115979.3 23 3945
XM_005253955.3 XP_005254012.1 23 4155
XM_005253956.3 XP_005254013.1 23 3752
XM_005253961.4 XP_005254018.1 12 2240
XM_011538867.2 XP_011537169.1 23 5207
XM_011538868.2 XP_011537170.1 23 5203
XM_011538869.2 XP_011537171.1 23 5205
XM_011538875.2 XP_011537177.1 12 2249
GenBank mRNA AB031230.1 - 14 2604
AJ459424.1 - 23 3917
AK027440.1 - 13 2334
AK027692.1 - 10 1850
AK074718.1 - 8 1429
AK127328.1 - 22 4041
AK299827.1 - 12 2259
AK316470.1 - 12 2252
AL133572.1 - 15 5740
BC008735.2 - 9 1715
BC115379.1 - 13 1935
BC115380.1 - 16 2175 40029132

View Full Product Details