Gene Transcripts
- Gene Symbol
- DARS
- Entrez Gene ID
- 1615
- Gene Name
- aspartyl-tRNA synthetase
- Gene Aliases
- HBSL, aspRS
- Chromosome Location
- Chr. 2: 135905881 - 135986046 on Build GRCh38
- UniGene ID
- Hs.503787
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001293312.1 |
NP_001280241.1
|
14, 15 |
1580 |
|
NM_001349.3 |
NP_001340.2
|
15, 16 |
1638 |
|
XM_017003489.1 |
XP_016858978.1
|
15, 16 |
1609 |
|
GenBank mRNA |
AK129521.1 |
- |
15, 16 |
1580 |
|
AK222476.1 |
- |
15, 16 |
1468 |
|
AK290607.1 |
- |
15, 16 |
1495 |
|
AY762100.1 |
- |
15, 16 |
1457 |
|
BC000629.2 |
- |
15, 16 |
1471 |
3344322 |
BC107749.1 |
- |
15, 16 |
1451 |
6709590 |
BT006710.1 |
- |
15, 16 |
1422 |
|
CR749809.1 |
- |
14, 15 |
1388 |
|
J05032.1 |
- |
15, 16 |
1512 |
|
View Full Product Details