Gene Transcripts

Gene Symbol
SPATA22
Entrez Gene ID
84690
Gene Name
spermatogenesis associated 22
Gene Aliases
NYD-SP20, NYDSP20
Chromosome Location
Chr. 17: 3440011 - 3513852 on Build GRCh38
UniGene ID
Hs.351068
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170695.1 NP_001164166.1 7 864
NM_001170696.1 NP_001164167.1 6 735
NM_001170697.1 NP_001164168.1 7 870
NM_001170698.1 NP_001164169.1 7 885
NM_001170699.1 NP_001164170.1 7 1104
NM_001321336.1 NP_001308265.1 7 1154
NM_001321337.1 NP_001308266.1 7 1202
NM_032598.4 NP_115987.2 7 926
GenBank mRNA AF367472.1 - 7 818
AK295299.1 - 7 1154
AK301892.1 - 7 1070
AY032684.1 - 6 684
AY035867.1 - 7 867
AY035868.1 - 7 926
BC029483.1 - 7 811 5271286

View Full Product Details