Gene Transcripts

Gene Symbol
PSRC1
Entrez Gene ID
84722
Gene Name
proline and serine rich coiled-coil 1
Gene Aliases
DDA3, FP3214
Chromosome Location
Chr. 1: 109279554 - 109283182 on Build GRCh38
UniGene ID
Hs.405925
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005290.3 NP_001005290.1 5 684
NM_001032291.2 NP_001027462.1 5 689
NM_032636.7 NP_116025.1 5 684
XM_005271283.2 XP_005271340.1 5 683
XM_011542306.2 XP_011540608.1 4 991
XM_017002560.1 XP_016858049.1 5 694
XM_017002561.1 XP_016858050.1 5 688
XM_017002562.1 XP_016858051.1 5 718
XM_017002563.1 XP_016858052.1 5 761
XM_017002564.1 XP_016858053.1 5 692
XM_017002565.1 XP_016858054.1 5 684
XM_017002566.1 XP_016858055.1 5 709
XM_017002567.1 XP_016858056.1 5 704
XM_017002568.1 XP_016858057.1 4 991
XM_017002569.1 XP_016858058.1 5 696
XM_017002570.1 XP_016858059.1 5 700
XM_017002571.1 XP_016858060.1 5 694
XM_017002572.1 XP_016858061.1 5 688
XM_017002573.1 XP_016858062.1 5 718
XM_017002574.1 XP_016858063.1 5 691
XM_017002575.1 XP_016858064.1 5 701
XM_017002576.1 XP_016858065.1 5 693
XM_017002577.1 XP_016858066.1 4 991
GenBank mRNA AF223000.1 - 5 663
AF447874.1 - 2 827
AK126567.1 - 5 620
AK289593.1 - 5 626
BC056909.1 - 5 702 6163125
BX649139.1 - 5 631

View Full Product Details