Gene Transcripts

Gene Symbol
PSRC1
Entrez Gene ID
84722
Gene Name
proline and serine rich coiled-coil 1
Gene Aliases
DDA3, FP3214
Chromosome Location
Chr. 1: 109279554 - 109283182 on Build GRCh38
UniGene ID
Hs.405925
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005290.3 NP_001005290.1 4 245
NM_001032291.2 NP_001027462.1 4 250
NM_032636.7 NP_116025.1 4 245
XM_005271283.2 XP_005271340.1 4 244
XM_011542306.2 XP_011540608.1 3 552
XM_017002560.1 XP_016858049.1 4 255
XM_017002561.1 XP_016858050.1 4 249
XM_017002562.1 XP_016858051.1 4 279
XM_017002563.1 XP_016858052.1 4 322
XM_017002564.1 XP_016858053.1 4 253
XM_017002565.1 XP_016858054.1 4 245
XM_017002566.1 XP_016858055.1 4 270
XM_017002567.1 XP_016858056.1 4 265
XM_017002568.1 XP_016858057.1 3 552
XM_017002569.1 XP_016858058.1 4 257
XM_017002570.1 XP_016858059.1 4 261
XM_017002571.1 XP_016858060.1 4 255
XM_017002572.1 XP_016858061.1 4 249
XM_017002573.1 XP_016858062.1 4 279
XM_017002574.1 XP_016858063.1 4 252
XM_017002575.1 XP_016858064.1 4 262
XM_017002576.1 XP_016858065.1 4 254
XM_017002577.1 XP_016858066.1 3 552
GenBank mRNA AF223000.1 - 4 224
AF447874.1 - 1 388
AK126567.1 - 4 181
AK289593.1 - 4 187
BC056909.1 - 4 263 6163125
BF984061.1 - 4 226
BJ992594.1 - 4 250
BX649139.1 - 4 192

View Full Product Details