Gene Transcripts

Gene Symbol
BRINP1
Entrez Gene ID
1620
Gene Name
BMP/retinoic acid inducible neural specific 1
Gene Aliases
DBC1, DBCCR1, FAM5A
Chromosome Location
Chr. 9: 119166630 - 119369461 on Build GRCh38
UniGene ID
Hs.532316
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014618.2 NP_055433.2 4 973
GenBank mRNA AF027734.1 - 4 935
BC021560.1 - 4 937
BC065196.1 - 4 926 5245480

View Full Product Details