Gene Transcripts

Gene Symbol
SFT2D3
Entrez Gene ID
84826
Gene Name
SFT2 domain containing 3
Gene Aliases
Chromosome Location
Chr. 2: 127701023 - 127703833 on Build GRCh38
UniGene ID
Hs.345849
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032740.3 NP_116129.3 1 1638
GenBank mRNA AK074549.1 - 1 1153
BC063637.1 - 1 106
BC110324.1 - 1 108

View Full Product Details