Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001144827.1 | NP_001138299.1 | 7 | 981 | |
NM_032795.2 | NP_116184.2 | 7 | 1074 | ||
XM_011543039.2 | XP_011541341.1 | 7 | 1071 | ||
GenBank mRNA | AK027400.1 | - | 7 | 1028 | |
AK225320.1 | - | 7 | 1034 | ||
AK300723.1 | - | 5 | 548 | ||
AK308059.1 | - | 7 | 934 | ||
BC014131.2 | - | 7 | 1035 | 4621434 | |
BE260782.1 | - | 1 | 72 |