Gene Transcripts

Gene Symbol
PPP1R15B
Entrez Gene ID
84919
Gene Name
protein phosphatase 1 regulatory subunit 15B
Gene Aliases
CREP, MSSGM2
Chromosome Location
Chr. 1: 204398684 - 204411880 on Build GRCh38
UniGene ID
Hs.304376
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032833.4 NP_116222.4 1 508
XM_005245551.4 XP_005245608.2 1 571
GenBank mRNA AK027650.1 - 1 508
AK222877.1 - 1 509
BC065280.1 - 1 482 6172811

View Full Product Details