Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001129889.2 | NP_001123361.1 | 6 | 1483 | |
NM_001322182.1 | NP_001309111.1 | 5 | 705 | ||
NM_001322183.1 | NP_001309112.1 | 6 | 759 | ||
NM_001322184.1 | NP_001309113.1 | 7 | 1059 | ||
NM_001322185.1 | NP_001309114.1 | 6 | 1005 | ||
NM_001322186.1 | NP_001309115.1 | 8 | 1274 | ||
NM_001922.4 | NP_001913.2 | 6 | 1483 | ||
XM_011521049.1 | XP_011519351.1 | 8 | 2107 | ||
XM_017020401.1 | XP_016875890.1 | 6 | 1486 | ||
GenBank mRNA | AK293115.1 | - | 6 | 1266 | |
BC028311.1 | - | 6 | 1330 | 4705935 | |
D17547.1 | - | 6 | 1470 | ||
DQ902581.1 | - | 6 | 1348 | ||
L18967.1 | - | 6 | 1440 | ||
S69231.1 | - | 6 | 1405 |