Gene Transcripts

Gene Symbol
MAEL
Entrez Gene ID
84944
Gene Name
maelstrom spermatogenic transposon silencer
Gene Aliases
CT128, SPATA35
Chromosome Location
Chr. 1: 166921121 - 167022214 on Build GRCh38
UniGene ID
Hs.651245
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286377.1 NP_001273306.1 9, 10 1223
NM_001286378.1 NP_001273307.1 11, 12 1089
NM_032858.2 NP_116247.1 10, 11 1316
XM_006711583.1 XP_006711646.1 11, 12 1120
XM_011510067.1 XP_011508369.1 10, 11 1282
XM_011510068.1 XP_011508370.1 11, 12 1073
XM_017002602.1 XP_016858091.1 12, 13 1457
XM_017002603.1 XP_016858092.1 11, 12 1119
GenBank mRNA AK027810.1 - 10, 11 1123
AK131066.1 - 10, 11 1126
AK302253.1 - 9, 10 1039
AL133073.1 - 9, 10 880
BC028595.2 - 10, 11 1096 5270252
BC034310.1 - 10, 11 1744
DQ076156.2 - 10, 11 1125
GU383113.1 - 9, 10 1032
GU383114.1 - 11, 12 1089

View Full Product Details