Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001281504.1 | NP_001268433.1 | 9 | 1454 | |
NM_032885.5 | NP_116274.3 | 9 | 1380 | ||
NR_104024.1 | 9 | 1336 | |||
NR_104025.1 | 8 | 1103 | |||
XM_006722927.1 | XP_006722990.1 | 7 | 1035 | ||
GenBank mRNA | AJ312332.1 | - | 9 | 1196 | |
AK056210.1 | - | 9 | 1355 | ||
AK223383.1 | - | 9 | 1305 | ||
AK294676.1 | - | 9 | 1454 | ||
AK314712.1 | - | 9 | 1315 | ||
BC007639.1 | - | 8 | 1026 | ||
BC016845.1 | - | 4 | 338 | ||
BC068992.1 | - | 9 | 1356 | 6578102 |