Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080593.2 | NP_542160.1 | 2 | 516 | |
GenBank mRNA | AJ223352.1 | - | 2 | 472 | |
BC000893.1 | - | 2 | 498 | 3463930 | |
BC051872.1 | - | 2 | 496 | 6526835 | |
BC064959.1 | - | 2 | 496 | 6093977 | |
BC108737.1 | - | 2 | 496 | 4707078 | |
DA873786.1 | - | 2 | 516 |