Gene Transcripts

Gene Symbol
KRTAP4-11
Entrez Gene ID
653240
Gene Name
keratin associated protein 4-11
Gene Aliases
KAP4.11, KAP4.14, KRTAP4-14, KRTAP4.14
Chromosome Location
Chr. 17: 41117181 - 41118354 on Build GRCh38
UniGene ID
Hs.307015
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033059.3 NP_149048.2 1 928
GenBank mRNA AJ406944.1 - 1 934
BC126131.1 - 1 910 8991847
BC130562.1 - 1 910 40146593

View Full Product Details